Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | ≥95.0% | 10000-60000 | 1 G
SDP
Catalog No. 5000404654
Encompass_Preferred
Poly(acrylic acid) is a synthetic polymer known for its versatility in various applications. It is often used as a superabsorbent, dispersant, thickener, and emulsifier in a range of industries.
  • Effective superabsorbent for liquids.
  • Acts as a dispersant in aqueous solutions.
  • Functions as a thickening agent.
  • Useful as an emulsifier.
  • Suitable for research use only.
Catalog No. 50-004-04654 Supplier Medchemexpress LLC Supplier No. HY4013831G
Poly(acrylic acid) is a synthetic polymer known for its versatility in various applications. It is often used as a superabsorbent, dispersant, thickener, and emulsifier in a range of industries.
  • Effective superabsorbent for liquids.
  • Acts as a dispersant in aqueous solutions.
  • Functions as a thickening agent.
  • Useful as an emulsifier.
  • Suitable for research use only.