Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | ≥95.0% | 10000-60000 | 500 MG
SDP
Catalog No. 5000404063
Encompass_Preferred
Poly(acrylic acid) (MW 10000-60000) is an anionic polymer. It can be utilized as a corrosion-mitigating and surface-stabilizing agent.
  • Anionic polymer
  • Corrosion-mitigating agent
  • Surface-stabilizing agent
Catalog No. 50-004-04063 Supplier Medchemexpress LLC Supplier No. HY401383500MG
Poly(acrylic acid) (MW 10000-60000) is an anionic polymer. It can be utilized as a corrosion-mitigating and surface-stabilizing agent.
  • Anionic polymer
  • Corrosion-mitigating agent
  • Surface-stabilizing agent