Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | MFCD00084394 | 3,000,000 (average) | (C3H4O2)x | 10 G
SDP
Catalog No. 5000280489
Encompass_Preferred
Poly(acrylic acid) (average MW 3,000,000) is a high-molecular-weight anionic polymer supplied as a solid for research use. It is commonly used in polymer science, surface treatment, corrosion inhibition studies, and biochemical assay formulations. Certificate of analysis and safety data sheet are available; follow the COA/SDS for handling, solubility, and storage instructions.
Catalog No. 50-002-80489 Supplier Medchemexpress LLC Supplier No. HYW243303B10G
Poly(acrylic acid) (average MW 3,000,000) is a high-molecular-weight anionic polymer supplied as a solid for research use. It is commonly used in polymer science, surface treatment, corrosion inhibition studies, and biochemical assay formulations. Certificate of analysis and safety data sheet are available; follow the COA/SDS for handling, solubility, and storage instructions.