Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | MFCD00084394 | 35.0% | 100,000 g/mol (average) | (C3H4O2)x | 10 ML
SDP
Catalog No. 5000282260
Encompass_Preferred
Poly(acrylic acid) (average Mw 100,000) is supplied as a 35% aqueous solution of an anionic polymer for research use. It is a colorless to light yellow liquid that is soluble in water and is used in biochemical and materials research where high-molecular-weight acrylic acid polymer solutions are required.
  • High average molecular weight suitable for viscosity and film-forming applications
  • Aqueous 35% formulation ready for laboratory use
  • Colorless to light yellow liquid for easy visual inspection
  • Store sealed at 4°C away from moisture to maintain stability
  • In solution: stable at -80°C for up to 6 months, or at -20°C for up to 1 month
  • Intended for research applications only
Catalog No. 50-002-82260 Supplier Medchemexpress LLC Supplier No. HYW243303M10ML
Poly(acrylic acid) (average Mw 100,000) is supplied as a 35% aqueous solution of an anionic polymer for research use. It is a colorless to light yellow liquid that is soluble in water and is used in biochemical and materials research where high-molecular-weight acrylic acid polymer solutions are required.
  • High average molecular weight suitable for viscosity and film-forming applications
  • Aqueous 35% formulation ready for laboratory use
  • Colorless to light yellow liquid for easy visual inspection
  • Store sealed at 4°C away from moisture to maintain stability
  • In solution: stable at -80°C for up to 6 months, or at -20°C for up to 1 month
  • Intended for research applications only