Learn More
Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | MFCD00084394 | 35.0% | 100000 (average) | (C3H4O2)x | 5 ML
SDP
Catalog No. 5000280763
Encompass_Preferred
Poly(acrylic acid) (Mv 100000) is provided as a 35% aqueous solution for research use. This high-molecular-weight anionic polymer is suitable for modifying viscosity, stabilizing dispersions, and altering surface properties in laboratory formulations. Supplied in small-volume vials for convenient handling and comes with a product datasheet and safety data sheet.
Catalog No. 50-002-80763 Supplier Medchemexpress LLC Supplier No. HYW243303M5ML
Poly(acrylic acid) (Mv 100000) is provided as a 35% aqueous solution for research use. This high-molecular-weight anionic polymer is suitable for modifying viscosity, stabilizing dispersions, and altering surface properties in laboratory formulations. Supplied in small-volume vials for convenient handling and comes with a product datasheet and safety data sheet.