Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Poly(acrylic acid) | 9003-01-4 | MFCD00084394 | 450,000 (average) | (C3H4O2)x | 5 G
SDP

Catalog No. 5000280760
Encompass_Preferred

Poly(acrylic acid) (average molecular weight 450,000) is a high-molecular-weight anionic polymer provided as a solid for use as a corrosion-mitigating and surface-stabilizing agent in biochemical and materials applications. The product is supplied in small research packs and includes manufacturer guidance for storage and handling.

  • Average molecular weight 450,000.
  • Anionic polymer used for corrosion mitigation and surface stabilization.
  • CAS number 9003-01-4.
  • Physical form: solid; store sealed and away from moisture.
  • Manufacturer provides data sheet, SDS, and COA for batch documentation.

Catalog No. 50-002-80760 Supplier Medchemexpress LLC Supplier No. HYW243303E5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(acrylic acid) (average molecular weight 450,000) is a high-molecular-weight anionic polymer provided as a solid for use as a corrosion-mitigating and surface-stabilizing agent in biochemical and materials applications. The product is supplied in small research packs and includes manufacturer guidance for storage and handling.

  • Average molecular weight 450,000.
  • Anionic polymer used for corrosion mitigation and surface stabilization.
  • CAS number 9003-01-4.
  • Physical form: solid; store sealed and away from moisture.
  • Manufacturer provides data sheet, SDS, and COA for batch documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.