Learn More
Medchemexpress LLC Poly(acrylic acid) (MW 10000~60000) | 9003-01-4 | ≥95.0% | 10000-60000 | 100 MG
SDP
Catalog No. 5000404804
Encompass_Preferred
Poly(acrylic acid) (MW 10000-60000) is a polyacrylic acid with a molecular weight of 10000-60000. It is an anionic polymer. It can be used as a corrosion-mitigating and surface-stabilizing agent.
  • Anionic polymer
  • Can be used as a corrosion-mitigating agent
  • Can be used as a surface-stabilizing agent
Catalog No. 50-004-04804 Supplier Medchemexpress LLC Supplier No. HY401383100MG
Poly(acrylic acid) (MW 10000-60000) is a polyacrylic acid with a molecular weight of 10000-60000. It is an anionic polymer. It can be used as a corrosion-mitigating and surface-stabilizing agent.
  • Anionic polymer
  • Can be used as a corrosion-mitigating agent
  • Can be used as a surface-stabilizing agent