Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(acrylic acid) partial sodium salt solution

Catalog No. 192023250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-202-3250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-202-3250G Supplier Sigma Aldrich Supplier No. 192023250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium salt of poly(acrylic acid) is a superabsorbent polymer. Superabsorbents are composed of crosslinked networks of hydrophilic polymers. It can absorb and retain a large capacity of water based fluids, the release of the absorbed fluids is tough even under an applied pressure. Absorption capacity is inversely proportional to salt concentration and applied load.They are formed via solution or inverse-suspension polymerization techniques. Sodium salt of poly (acrylic acid) may be used as a major component of non glare coatings for headlights.It acts as an antiscaling agents.It's effect of the salt on the crystal (Cu and MS metals) formation was studied. General applications of these superabsorbents are in many fields such as agriculture and horticulture, sanitary goods, sealing composites, component of moisture barrier tapes and coatings, humidity control, gel actuators. and medicine for drug-delivery systems.

Specifications

CAS 9033-79-8
Density 1.2 g/mL (at 25°C)
Refractive Index n20/D 1.442
Quantity 250 g
MDL Number MFCD00084394
Synonym Acrylic acid-sodium acrylate copolymer; Sodium acrylate-acrylic acid copolymer
Molecular Weight (g/mol) 2000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.