Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(acrylonitrile-co-butadiene), dicarboxy terminated

Catalog No. 418870250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-887-0250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-887-0250ML Supplier Sigma Aldrich Supplier No. 418870250ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Component of epoxy-based adhesives, composites and coatings. Intermediate in corrosion-resistant vinyl esters, acrylated epoxies used in radiation curing and photopolymer printing plates.

Specifications

CAS 68891-46-3
Density 0.924 g/mL (at 25°C)
Boiling Point specific activity: 1.9 carboxyl groups / molecule
Refractive Index n20/D 1.515 (literature)
Linear Formula HO2C[CH2CH(CN)]x(CH2CH=CHCH2)yCO2H
MDL Number MFCD06591088
Quantity 250 mL
Molecular Weight (g/mol) 3800
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.