Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(acrylonitrile-co-methyl acrylate)

Catalog No. 51794110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-794-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-794-110G Supplier Sigma Aldrich Supplier No. 51794110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(acrylonitrile-co-methyl acrylate) (P(AN-co-MA)) may be used to fabricate multilayer mixed matrix membranes (MMMMs) for easy separation of ethanol from an ethanol-water mixture.

Specifications

CAS 24968-79-4
Linear Formula [CH2CH(CN)]x[CH2CH(CO2CH3)]y
MDL Number MFCD00284236
Quantity 10 g
Synonym Acrylic acid methyl ester, polymer with acrylonitrile; Acrylonitrile-methyl acrylate copolymer; Acrylonitrile-methyl acrylate polymer; Methyl acrylate-acrylonitrile copolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.