Learn More
Enzo Life Sciences Poly(ADP-ribose) monoclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2160100
Encompass_Preferred
Clone: not available. Host: Mouse. Isotype: IgG3. Immunogen: Purified poly(ADP-ribose) polymer (chain length of 2-50 units). Species Reactivity: Species independent. Applications: ELISA, ICC, IP, WB. Recommended dilutions: Western Blot (1:1,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Purified from mouse ascites. Formulation: Liquid. In PBS containing 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. NC1633719