Learn More
Cusabio Technology LLC poly ADP-ribose polymerase 1
Catalog No. 5000049265
Encompass_Preferred
The Human Poly ADP Ribose Polymerase (PARP) ELISA Kit is designed for the quantitative detection of PARP1 in serum plasma tissue homogenates and cell lysates This kit detects PARP1 using a sensitive sandwich ELISA method with a detection range of 15 6 pg/mL to 1000 pg/mL and a minimum detectable concentration of 3 9 pg/mL The assay requires a sample volume of 50-100 L and employs a 1-5 hour incubation period for optimal results Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable quantification of PARP1 levels in various biological samples This kit is suitable for research applications requiring precise assessment of PARP1 expression in studies related to DNA repair apoptosis and cellular stress responses
Catalog No. 50-000-49265 Supplier Cusabio Technology LLC Supplier No. CSBE10705H
The Human Poly ADP Ribose Polymerase (PARP) ELISA Kit is designed for the quantitative detection of PARP1 in serum plasma tissue homogenates and cell lysates This kit detects PARP1 using a sensitive sandwich ELISA method with a detection range of 15 6 pg/mL to 1000 pg/mL and a minimum detectable concentration of 3 9 pg/mL The assay requires a sample volume of 50-100 L and employs a 1-5 hour incubation period for optimal results Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable quantification of PARP1 levels in various biological samples This kit is suitable for research applications requiring precise assessment of PARP1 expression in studies related to DNA repair apoptosis and cellular stress responses