Learn More
Biomatik Corporation Poly Adp Ribose Polymerase (Parp) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU30614
Encompass_Preferred
Poly Adp Ribose Polymerase (Parp) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Poly ADP Ribose Polymerase (PARP). The Poly Adp Ribose Polymerase (Parp) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp788~Trp1014. Applications: WB, IHC, ICC, IP. Target Synonyms: Poly ADP Ribose Polymerase (PARP). Accession Number: P09874. Gene ID: 142. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5011