Learn More
Enzo Life Sciences Poly(ADP-ribose), standard (1µg)
SDP
Catalog No. 502008047
Encompass_Preferred
Alternative name: PAR. Purity: ≥95% (HPLC of ADP-ribose RNA and DNA measurement). MW: 1’100 to 275’000 depending on the number of units of ADP-ribose. Formulation: Liquid. In water. Source: Released from automodified PARP-1 by base treatment and purified by boronate affinity. Long Term Storage: -80°C. Use/Stability: Stable for one week when stored at +4°C. Stable for at least 2 years after receipt when stored at -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8047 Supplier Enzo Life Sciences Supplier No. ALX202043C001
Alternative name: PAR. Purity: ≥95% (HPLC of ADP-ribose RNA and DNA measurement). MW: 1’100 to 275’000 depending on the number of units of ADP-ribose. Formulation: Liquid. In water. Source: Released from automodified PARP-1 by base treatment and purified by boronate affinity. Long Term Storage: -80°C. Use/Stability: Stable for one week when stored at +4°C. Stable for at least 2 years after receipt when stored at -80°C. Handling: Avoid freeze/thaw cycles.