Learn More
Uline 1 Mil Poly Bags, Clear, 500
Catalog No. NC2326146
Encompass_Preferred
Lightweight protection for temporary storage, 100 virgin high clarity polyethylene film, Meets FDA and USDA specifications for food contact, Low/Linear Low Density, Sulfur free, Amine and Silicone free, Latex free, BHT free Butylated Hydroxy Toluene, Chlorine free, PVC free, Temperature range 20 - 150 F, Recyclable 4, Bottom seal, Not treated for printing by end-user, Not recommended for dry ice, Not microwaveable, Cannot be used for baking cooking or boiling, Do not store in sub-freezing temperatures, 3 or 4 mil is recommended for use in a trash compactor, Water-resistant not waterproof, Cannot use with vacuum machine, Meets FDA 21 CFR 177.1520, Acid free and archival safe
Catalog No. NC2326146 Supplier Uline Supplier No. S10913
Lightweight protection for temporary storage, 100 virgin high clarity polyethylene film, Meets FDA and USDA specifications for food contact, Low/Linear Low Density, Sulfur free, Amine and Silicone free, Latex free, BHT free Butylated Hydroxy Toluene, Chlorine free, PVC free, Temperature range 20 - 150 F, Recyclable 4, Bottom seal, Not treated for printing by end-user, Not recommended for dry ice, Not microwaveable, Cannot be used for baking cooking or boiling, Do not store in sub-freezing temperatures, 3 or 4 mil is recommended for use in a trash compactor, Water-resistant not waterproof, Cannot use with vacuum machine, Meets FDA 21 CFR 177.1520, Acid free and archival safe