Learn More
MSC Value Collection Polybags, 3 x 5 in, 1.5 Mil, 1000/Pack
Catalog No. 501917114
Encompass
Value Collection polybags are clear, reclosable plastic bags designed for secure storage and enhanced product presentation. They are suitable for various applications requiring moisture, grease, and vapor resistance.
  • Width of 3 in. and a height of 5 in.
  • Thickness of 1.5 mil
  • Made from polypropylene material
  • Resealable design with easy peel-and-fold closure
  • Resistant to moisture, grease, and vapor
  • Complies with FDA and USDA specifications for food contact
  • High clarity enhances visual appeal of contents
  • Dimensions represent inside measurements
  • Sold in packages containing 1000 units
Catalog No. 50-191-7114 Supplier MSC Supplier No. 39566633
Value Collection polybags are clear, reclosable plastic bags designed for secure storage and enhanced product presentation. They are suitable for various applications requiring moisture, grease, and vapor resistance.
  • Width of 3 in. and a height of 5 in.
  • Thickness of 1.5 mil
  • Made from polypropylene material
  • Resealable design with easy peel-and-fold closure
  • Resistant to moisture, grease, and vapor
  • Complies with FDA and USDA specifications for food contact
  • High clarity enhances visual appeal of contents
  • Dimensions represent inside measurements
  • Sold in packages containing 1000 units