Learn More
MSC POLY BLEND C/R GLV
Catalog No. 5000675411
Encompass
These gloves are designed for cut, puncture, and abrasion resistance, featuring RhinoYarn material with a polyurethane coating on the palm and fingertips for a smooth grip.
  • Knit style
  • Polyurethane coating material
  • Palm and fingertips coating coverage
  • ANSI/ISEA cut resistance level A4
  • Primary material RhinoYarn
  • ANSI/ISEA abrasion resistance level 3
  • Lime color
  • Large glove size (numeric size 9)
  • ANSI/ISEA puncture resistance level 3
  • Black polyurethane coated palm
  • EN 388 rating 3X42D
  • RhinoYarn back material
  • Not FDA approved
  • Seamless RhinoYarn lining
  • No blood borne pathogen barrier
  • Unisex gender
Catalog No. 50-006-75411 Supplier MSC Supplier No. 90691916
These gloves are designed for cut, puncture, and abrasion resistance, featuring RhinoYarn material with a polyurethane coating on the palm and fingertips for a smooth grip.
  • Knit style
  • Polyurethane coating material
  • Palm and fingertips coating coverage
  • ANSI/ISEA cut resistance level A4
  • Primary material RhinoYarn
  • ANSI/ISEA abrasion resistance level 3
  • Lime color
  • Large glove size (numeric size 9)
  • ANSI/ISEA puncture resistance level 3
  • Black polyurethane coated palm
  • EN 388 rating 3X42D
  • RhinoYarn back material
  • Not FDA approved
  • Seamless RhinoYarn lining
  • No blood borne pathogen barrier
  • Unisex gender