Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(butadiene), cis and trans, Thermo Scientific™

Catalog Number AC178131000
Click to view available options
Quantity:
100 g
9003-17-2
C4H6
54.092
KAKZBPTYRLMSJV-UHFFFAOYSA-N
1,3-butadiene, divinyl, butadiene, vinylethylene, biethylene, erythrene, bivinyl, pyrrolylene, butadieen, butadien
7845
CHEBI:39478
buta-1,3-diene
C=CC=C
9003-17-2
C4H6
54.092
KAKZBPTYRLMSJV-UHFFFAOYSA-N
1,3-butadiene, divinyl, butadiene, vinylethylene, biethylene, erythrene, bivinyl, pyrrolylene, butadieen, butadien
7845
CHEBI:39478
buta-1,3-diene
C=CC=C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Slab
Linear Formula (-CH2CH2CH=CH-)n
Density 0.9g/mL
Quantity 100 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Chemical Name or Material Poly(butadiene)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit