Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(butyl acrylate) solution

Catalog No. 18140450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-140-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-140-450G Supplier Sigma Aldrich Supplier No. 18140450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Unit weight does not include weight of solvent.

Specifications

CAS 9003-49-0
Density 1.087 g/mL (at 25°C)
Linear Formula (C7H12O2)n
Refractive Index n20/D 1.466 (literature)
MDL Number MFCD00084401
Quantity 50 g
Synonym Acrylic acid butyl ester polymer; Butyl acrylate homopolymer; Butyl acrylate polymer; Poly(n -butyl acrylate)
Molecular Weight (g/mol) 99000
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.