Learn More
Sigma Aldrich Poly(caprolactone-co-glycolide)
Catalog No. 9010075G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
Catalog No. Quantity
90-100-75G 5 g
1 options
Catalog No. 90-100-75G Supplier Sigma Aldrich Supplier No. 9010075G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Poly(caprolactone-co-glycolide) (PGCL) is a biodegradable and biocompatible copolymer synthesized from the ring opening polymerization of glycolide and epsilon-caprolactone. Due to the ester linkages found in the polymer backbone, these copolymers are able to degrade via hydrolysis. Depending on the ratio of monomers used and polymer block structure, different forms of PGCL can be obtained. These differing forms yield materials with different degrees of crystallinity, thermal transitions, solubility, and rates of degradation.Poly(caprolactone-co-glycolide) (PGCL) is a biodegradable and biocompatible copolymer for use in biomedical research devices, such as implants, sutures, prosthetic devices, scaffolds for tissue engineering applications, micro and nanoparticles. PGA-PCL is also utilized in drug delivery research, such as in the synthesis and application of polymer-drug conjugates.
Specifications
Linear Formula (C4H4O4)m(C6H10O2)n
Quantity 5 g