Learn More
MSC Pro-Safe Poly Carpet Entrance Mat, Moss Green, Ribbed, 4 ft. x 8 ft., 1 ea
Catalog No. 5000688008
Encompass
This moss green entrance mat features a durable ribbed poly-blended carpet surface designed for effective dirt and moisture removal in indoor areas with medium traffic. The construction includes a vinyl base for added stability and reliable performance in demanding environments.
  • Ribbed surface pattern for scraping debris
  • Poly-blended carpet material
  • Vinyl base construction
  • Designed for indoor use
  • Suitable for medium traffic environments
  • Dimensions: 8 ft. long x 4 ft. wide
  • Thickness: 5/16 in.
Catalog No. 50-006-88008 Supplier MSC Supplier No. 88266044
This moss green entrance mat features a durable ribbed poly-blended carpet surface designed for effective dirt and moisture removal in indoor areas with medium traffic. The construction includes a vinyl base for added stability and reliable performance in demanding environments.
  • Ribbed surface pattern for scraping debris
  • Poly-blended carpet material
  • Vinyl base construction
  • Designed for indoor use
  • Suitable for medium traffic environments
  • Dimensions: 8 ft. long x 4 ft. wide
  • Thickness: 5/16 in.