Learn More
Research Products International Corp Graduated Centrifuge Tubes, 50ml, Polypropylene, Sterile, 50 per Bag, 500 per Case
SDP
Catalog No. 502114079
Encompass
Premium quality centrifuge tubes are available in a 50ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -40°C to +125°C and can be centrifuged at maximum of 10,000 RCF(g).
Catalog No. 50-211-4079 Supplier Research Products International Corp Supplier No. 163228LC
Premium quality centrifuge tubes are available in a 50ml size. High clarity polypropylene has excellent chemical and temperature resistance. Tubes withstand temperature extremes from -40°C to +125°C and can be centrifuged at maximum of 10,000 RCF(g).