Learn More
Bevco Precision Manufacturing Co Dura Desk Height Heavy-Duty ISO 5 Cleanroom Black Polyurethane Chair; Articulating Seat & Back Tilt; Black Nylon Base
Catalog No. NC2023060 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
BEVCO's Dura ISO 5 Cleanroom heavy-duty polyurethane chairs are designed to meet the particulate goals of ISO 14644-1 and have been tested by an independent laboratory to certify ISO 5 compliance. The black polyurethane seat and back are impervious to water, oil, and most chemicals and are puncture resistant and easy to clean. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. The contoured back is fully adjustable encouraging proper spinal alignment. The large waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 15" - 20". This chair comes with a sturdy 25" diameter black nylon base. Model comes standard with mushroom glides. Optional casters can be ordered separately.
Catalog No. NC2023060 Supplier Bevco Precision Manufacturing Co Supplier No. 7001DC
BEVCO's Dura ISO 5 Cleanroom heavy-duty polyurethane chairs are designed to meet the particulate goals of ISO 14644-1 and have been tested by an independent laboratory to certify ISO 5 compliance. The black polyurethane seat and back are impervious to water, oil, and most chemicals and are puncture resistant and easy to clean. This articulating tilt model features simultaneous seat and back adjustments to properly position the angle of the chair for increased comfort during extended sitting durations. The contoured back is fully adjustable encouraging proper spinal alignment. The large waterfall seat aids in lower body circulation, improving overall wellness. The pneumatic height adjustment allows you to easily change your seat height from 15" - 20". This chair comes with a sturdy 25" diameter black nylon base. Model comes standard with mushroom glides. Optional casters can be ordered separately.