Learn More
Electron Microscopy Sciences Poly-Container Poly-Container 3 Dram, 11 ml
SDP
Catalog No. 5028023
Encompass_Preferred
Lightweight, air-tight containers with snap-on polyethylene caps. They have less resistance to breakage than polyethylene but they are transparent and are ideal when used as a specimen container, storage vessel, or a block holder.
Volume: 11 ml
Diameter: 19 mm
Height: 48 mm
Catalog No. 50-280-23 Supplier Electron Microscopy Sciences Supplier No. 6424810
Add to Cart
Add to Cart
Lightweight, air-tight containers with snap-on polyethylene caps. They have less resistance to breakage than polyethylene but they are transparent and are ideal when used as a specimen container, storage vessel, or a block holder.
Volume: 11 ml
Diameter: 19 mm
Height: 48 mm