Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(cyclohexyl methacrylate)

Catalog No. 19194925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-194-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-194-925G Supplier Sigma Aldrich Supplier No. 19194925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The four relaxation process of poly(cyclohexyl methacrylate) was studied by investigating its viscoelastic and dielectric properties over a range of temperatures.Poly(cyclohexyl methacrylate (PCHMA) may be used to prepare polymeric blends with polystyrene and nanocomposites with carbon nanotubes. Densely grafted PCHMA on multiwalled carbon nanotubes (MWCNTs) composites exhibits better mechanical properties and electrical resistance compared to PCHMA blended CNTs composites.

Specifications

CAS 25768-50-7
Density 1.1 g/mL (at 25°C)
Refractive Index n20/D 1.5065 (literature)
Linear Formula (C10H16O2)n
Quantity 25 g
MDL Number MFCD00084408
Synonym PCMA
Molecular Weight (g/mol) 65000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.