Learn More
AdipoGen Poly-D-galacturonic acid ME
SDP
Catalog No. 502560201
Encompass_Preferred
Chemical. CAS 9000-69-5. Formula . MW . Citrus peel. Pectin is a structural heteropolysaccharide found ubiquitously in terrestrial plants. It finds diverse food applications such as that of a gelling agent, stabilizer and fat replacer. Used as a substrate to identify, differentiate and characterize pectinases and it is used to study its degradation by pectinolytic bacteria. It acts as anti-metastatic, cholesterol inhibiting, prebiotic and probiotic encapsulant. Pectin is being widely used as a component of bio-nano-packaging.
Catalog No. 50-256-0201 Supplier AdipoGen Supplier No. CDXP0233G250
Add to Cart
Add to Cart
Chemical. CAS 9000-69-5. Formula . MW . Citrus peel. Pectin is a structural heteropolysaccharide found ubiquitously in terrestrial plants. It finds diverse food applications such as that of a gelling agent, stabilizer and fat replacer. Used as a substrate to identify, differentiate and characterize pectinases and it is used to study its degradation by pectinolytic bacteria. It acts as anti-metastatic, cholesterol inhibiting, prebiotic and probiotic encapsulant. Pectin is being widely used as a component of bio-nano-packaging.