Learn More
Polysciences, Inc. Poly(D,L-lactic acid), IV 0.4 dl/g26680-10-4Molecular Weight: 20,000 - 30,000
SDP
Catalog No. NC1749739
Encompass_Preferred
Poly(D,L-lactic acid), IV 0.4 dl/g26680-10-4Molecular Weight: 20,000 - 30,000
Catalog No. NC1749739 Supplier Polysciences, Inc. Supplier No. 1658510
Poly(D,L-lactic acid), IV 0.4 dl/g26680-10-4Molecular Weight: 20,000 - 30,000