Learn More
Polysciences, Inc. POLY(D,L-LACTIDE-CO-10G
SDP
Catalog No. NC2294445
Encompass_Preferred
Poly(D,L-lactide-co-glycolide), 50:50, IV 0.40 dl/g, 10G
This item is not returnable. View return policy
Catalog No. NC2294445 Supplier Polysciences, Inc. Supplier No. 2629710
Poly(D,L-lactide-co-glycolide), 50:50, IV 0.40 dl/g, 10G