Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(D,L-lactide-co-glycolide)

Catalog No. 4304715G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-047-15G 5 g
43-047-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-047-15G Supplier Sigma Aldrich Supplier No. 4304715G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Characteristic properties of ester terminated poly(D,L-lactide-co-glycolide) (PLGA) are in vivo biodegradability, tunable drug release profile and very high encapsulation capacity.PLGA polymers find uses in the production of biodegradable medical sutures and for controlled drug release. PLGA microspheres have been reported for use in controlled release of human growth hormone. Thin membranes of blended polymers constituting of PLGA/polycaprolactone/α-tricalcium phosphate have been used in guided bone regeneration.Lactel is a registered trademark of Durect Corp.

Specifications

Linear Formula [C3H4O2]x[C2H2O2]y
MDL Number MFCD00131930
Quantity 5 g
Synonym Lactel™ B6006-1; PLGA; PLGA
Molecular Weight (g/mol) 50,000-75,000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.