Learn More
Medchemexpress LLC Poly(D,L-lactide-co-glycolide) | 26780-50-7 | MFCD00131930 | 260.2 g/mol | (C6H8O4.C4H4O4)x | 500mg
SDP
Catalog No. 5000270396
Encompass_Preferred
Poly(D L-lactide-co-glycolide) is a synthetic polymer material with good biocompatibility Poly(D L-lactide-co-glycolide) can be used in the research of drug delivery systems[1]
Catalog No. 50-002-70396 Supplier Medchemexpress LLC Supplier No. HYW723152500MG
Poly(D L-lactide-co-glycolide) is a synthetic polymer material with good biocompatibility Poly(D L-lactide-co-glycolide) can be used in the research of drug delivery systems[1]