Learn More
Medchemexpress LLC Poly-D-lysine hydrobromide | 27964-99-4 | MFCD00131934 | 96.0% | C22H51BrN6O3 | 10MG
SDP
Catalog No. 5000260569
Encompass_Preferred
Poly-D-lysine hydrobromide (average MW 84,000) is a polycationic polymer used to promote cell adhesion on culture surfaces, particularly for neuronal and mammalian cell cultures. It is supplied as a white to off-white solid and is water-soluble with ultrasonic assistance, making it suitable for preparing coating solutions to improve cell attachment and spreading.
  • Promotes cell adhesion for neural and mammalian cultures.
  • Polycationic nature enhances substrate binding and stimulates endocytosis.
  • High average molecular weight supports formation of stable surface coatings.
  • Soluble in water (requires ultrasonic) for convenient solution preparation.
  • Available in small research-sized pack sizes for laboratory use.
  • Stable when stored sealed; recommended cold storage to preserve integrity.
Catalog No. 50-002-60569 Supplier Medchemexpress LLC Supplier No. HY139201B10MG
Poly-D-lysine hydrobromide (average MW 84,000) is a polycationic polymer used to promote cell adhesion on culture surfaces, particularly for neuronal and mammalian cell cultures. It is supplied as a white to off-white solid and is water-soluble with ultrasonic assistance, making it suitable for preparing coating solutions to improve cell attachment and spreading.
  • Promotes cell adhesion for neural and mammalian cultures.
  • Polycationic nature enhances substrate binding and stimulates endocytosis.
  • High average molecular weight supports formation of stable surface coatings.
  • Soluble in water (requires ultrasonic) for convenient solution preparation.
  • Available in small research-sized pack sizes for laboratory use.
  • Stable when stored sealed; recommended cold storage to preserve integrity.