Learn More
Sigma Aldrich Fine Chemicals Biosciences Poly-D-lysine hydrobromide, 27964-99-4, MFCD00131934
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P64075MG
Encompass
Linear Formula D-Lys-(D-Lys)n-D-Lys · xHBr,mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture. Poly-D-lysine polymers can be used in preparing surfaces for cell attachment. The D-lysine polymers can also be used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.
Catalog No. 50-165-6955