Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-D-Lysine Hydrobromide, MP Biomedicals

Catalog No. ICN10269480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
1 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10269480 100 mg
ICN10269410 10 mg
ICN10269450 50 mg
ICN10269490 500 mg
ICN10269491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10269480 Supplier MP Biomedicals, Inc Supplier No. 0210269480
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27964-99-4
Molecular Formula C6H15BrN2O2
Molecular Weight (g/mol) 227.102
InChI Key MEXAGTSTSPYCEP-NUBCRITNSA-N
Synonym d-lysine hydrobromide
PubChem CID 87493163
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrobromide
SMILES C(CCN)CC(C(=O)O)N.Br

Specifications

Quantity 100 mg
Formula Weight 4,000 to 15,000
Chemical Name or Material Poly-D-Lysine Hydrobromide
Recommended Storage 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.