Learn More
Invivogen dsDNA Naked, Complexed with Transfection Reagent, 100 µg
SDP
Catalog No. NC0119678
Encompass
Poly(dA:dT) is a poly(deoxyadenylic-deoxythymidylic) acid sodium salt. It is a repetitive synthetic double-stranded DNA sequence of poly(dA-dT); poly(dT-dA) and a synthetic analog of B-DNA. Poly(dA:dT) is recognized by several sensors, including DAI, LRRFIP1 and AIM2. Poly(dA:dT) is also indirectly detected by RIG-I. The activity of poly(dA:dT) is tested using the reporter cell line B16-Blue IFN/. Solubility: 2 mg/ml in physiological water. Unit Size:100 g. 86828-69-5
Catalog No. NC0119678 Supplier Invivogen Supplier No. TLRLPATC
Poly(dA:dT) is a poly(deoxyadenylic-deoxythymidylic) acid sodium salt. It is a repetitive synthetic double-stranded DNA sequence of poly(dA-dT); poly(dT-dA) and a synthetic analog of B-DNA. Poly(dA:dT) is recognized by several sensors, including DAI, LRRFIP1 and AIM2. Poly(dA:dT) is also indirectly detected by RIG-I. The activity of poly(dA:dT) is tested using the reporter cell line B16-Blue IFN/. Solubility: 2 mg/ml in physiological water. Unit Size:100 g. 86828-69-5