Learn More
Invivogen Poly(dA:dT) dsDNA naked, 1 mg
SDP
Catalog No. NC0087992
Encompass
Poly(dA:dT) is a poly(deoxyadenylic-deoxythymidylic) acid sodium salt. It is a repetitive synthetic double-stranded DNA sequence of poly(dA-dT):poly(dT-dA) and a synthetic analog of B-DNA.
Catalog No. NC0087992 Supplier Invivogen Supplier No. TLRLPATN1
Poly(dA:dT) is a poly(deoxyadenylic-deoxythymidylic) acid sodium salt. It is a repetitive synthetic double-stranded DNA sequence of poly(dA-dT):poly(dT-dA) and a synthetic analog of B-DNA.