Learn More
Akina Inc Poly(DL-lactide) (per g)
SDP
Catalog No. NC1576990
Encompass
PDLLA (Mn 25,000 - 35,000 Da) acid endcap DL Poly(DL-lactide) (per g)
Catalog No. NC1576990 Supplier Akina Inc Supplier No. AP2241G
PDLLA (Mn 25,000 - 35,000 Da) acid endcap DL Poly(DL-lactide) (per g)