Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethyl methacrylate)

Catalog No. 182087250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-208-7250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-208-7250G Supplier Sigma Aldrich Supplier No. 182087250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PEMA is known to exhibit biocompatibility and in vivo stability in its various use in implants and medical devices. Sorption of low solubility gases in PEMA was observed to be comparatively lower that polymers with high transition temperature.Poly(ethyl methacrylate) (PEMA) may be used to prepare polymer blends with polystyrene using organically modified clay as emulsifier.

Specifications

CAS 9003-42-3
Density 1.11 g/mL (at 25°C)
Refractive Index n20/D 1.485
Linear Formula [CH2C(CH3)(CO2C2H5)]n
Quantity 250 g
MDL Number MFCD00084414
Synonym Ethyl methacrylate homopolymer; Poly(ethyl 2-methyl-2-propeonate)
Molecular Weight (g/mol) 515000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.