Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene-co-methyl acrylate-co-glycidyl methacrylate)

Catalog No. 433640250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-364-0250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-364-0250G Supplier Sigma Aldrich Supplier No. 433640250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene-co-methyl acrylate-co-glycidyl methacrylate)

Specifications

CAS 51541-08-3
Melting Point 39°C
Density 0.94 g/mL (at 25°C)
MDL Number MFCD00217719
Quantity 250 g
Synonym Ethylene-methyl acrylate-glycidyl methacrylate copolymer; Glycidyl methacrylate-ethylene-methyl acrylate copolymer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.