Learn More
Medchemexpress LLC Poly ethylene glycol 100mg
SDP
Catalog No. 5000204892
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 3500) (PEGDA (MW 3500)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 3500) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-04892 Supplier Medchemexpress LLC Supplier No. HYW110542L100MG
Poly(ethylene glycol) diacrylate (MW 3500) (PEGDA (MW 3500)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 3500) can be used in the formation of a cross-linked polymers[1]