Learn More
Medchemexpress LLC Poly ethylene glycol 100mg
SDP
Catalog No. 5000205467
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 20000) (PEGDA (MW 20000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 20000) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-05467 Supplier Medchemexpress LLC Supplier No. HYW110542M100MG
Poly(ethylene glycol) diacrylate (MW 20000) (PEGDA (MW 20000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 20000) can be used in the formation of a cross-linked polymers[1]