Learn More
Medchemexpress LLC Poly ethylene glycol 100mg
SDP
Catalog No. 5000204893
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor (PEGDA (MW 4000) MEHQ as inhibitor) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-04893 Supplier Medchemexpress LLC Supplier No. HYW110542P100MG
Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor (PEGDA (MW 4000) MEHQ as inhibitor) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor can be used in the formation of a cross-linked polymers[1]