Learn More
Sigma Aldrich Poly(ethylene glycol) (12) tridecyl ether
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 78330-21-9 |
| Density | 1 g/mL (at 25°C) |
| Boiling Point | >150°C |
| Refractive Index | n20/D 1.461 (literature) |
| Linear Formula | C13H27(OCH2CH2)nOH, n ∼12 |
| Quantity | 100 mL |
| MDL Number | MFCD00132049 |
| Synonym | Polyoxyethylene (12) tridecyl ether |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.