Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) (12) tridecyl ether

Catalog No. 46641725L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-641-725L 2.5 L
46-641-7100ML 100 mL
46-641-7500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 46-641-725L Supplier Sigma Aldrich Supplier No. 4664172.5L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to enhance conductivity of poly(3,4-ethyledioxythiopene)PEDOT:poly (styrene sulfonate)PSS, next generation transperant electrode materials for organic solar cells.It is also majorly used as a surfactant.

Specifications

CAS 78330-21-9
Density 1 g/mL (at 25°C)
Boiling Point >150°C
Refractive Index n20/D 1.461 (literature)
Linear Formula C13H27(OCH2CH2)nOH, n ∼12
MDL Number MFCD00132049
Quantity 2.5 L
Synonym Polyoxyethylene (12) tridecyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.