Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) (18) tridecyl ether

Catalog No. 466875100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-687-5100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 46-687-5100G Supplier Sigma Aldrich Supplier No. 466875100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) (18) tridecyl ether

Specifications

CAS 24938-91-8
Melting Point 28°C
Density 0.96 g/mL (at 25°C)
Refractive Index n20/D 1.464 (literature)
Linear Formula C13H27(OCH2CH2)nOH, n ~18
MDL Number MFCD00132049
Quantity 100 g
Synonym Polyoxyethylene (18) tridecyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.