Learn More
Medchemexpress LLC Poly ethylene glycol 1g
SDP
Catalog No. 5000205499
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 6000) (PEGDA (MW 6000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 6000) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-05499 Supplier Medchemexpress LLC Supplier No. HYW110542G1G
Poly(ethylene glycol) diacrylate (MW 6000) (PEGDA (MW 6000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 6000) can be used in the formation of a cross-linked polymers[1]