Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) 2-mercaptoethyl ether acetic acid

Catalog No. 757837500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-783-7500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-783-7500MG Supplier Sigma Aldrich Supplier No. 757837500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reported applications include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization.

Specifications

CAS 165729-81-7
Linear Formula HSC2H4O(C2H4O)nCH2CO2H
Quantity 500 mg
Synonym mercaptoPEG acid; mercaptopoly(ethylene glycol) carboxylic acid; mercaptopolyoxyethylene acetic acid; thio-PEG; thiol-PEG-acid; thiol-PEG-carboxylate
Molecular Weight (g/mol) 340077
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.