Learn More
Medchemexpress LLC Poly ethylene glycol 250mg
SDP
Catalog No. 5000205768
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 1000) (PEGDA (MW 1000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 1000) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-05768 Supplier Medchemexpress LLC Supplier No. 250MGHYW110542D
Poly(ethylene glycol) diacrylate (MW 1000) (PEGDA (MW 1000)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 1000) can be used in the formation of a cross-linked polymers[1]