Learn More
Medchemexpress LLC Poly ethylene glycol 250mg
SDP
Catalog No. 5000206017
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor (PEGDA (MW 4000) MEHQ as inhibitor) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-06017 Supplier Medchemexpress LLC Supplier No. 250MGHYW110542P
Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor (PEGDA (MW 4000) MEHQ as inhibitor) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 4000) MEHQ as inhibitor can be used in the formation of a cross-linked polymers[1]