Learn More
Medchemexpress LLC Poly ethylene glycol 250uL
SDP
Catalog No. 5000205312
Encompass_Preferred
Poly(ethylene glycol) diacrylate (MW 700) (PEGDA (MW 700)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 700) can be used in the formation of a cross-linked polymers[1]
Catalog No. 50-002-05312 Supplier Medchemexpress LLC Supplier No. 250ULHYW110542J
Poly(ethylene glycol) diacrylate (MW 700) (PEGDA (MW 700)) is a derivative of polyethylene glycol Poly(ethylene glycol) diacrylate (MW 700) can be used in the formation of a cross-linked polymers[1]