Learn More
Description
Specifications
Specifications
| CAS | 25322-68-3 |
| Melting Point | 4°C to 8°C |
| Density | 1.128 g/mL |
| Linear Formula | H(OCH2CH2)nOH |
| Refractive Index | n20/D 1.466 |
| MDL Number | MFCD00081839 |
| Quantity | 5 g |
| Synonym | PEG |
| Molecular Weight (g/mol) | 380 to 420 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.